Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhp2
Entrez Gene ID
100003787
Gene Name
NHP2 ribonucleoprotein homolog (yeast)
Gene Aliases
nola2, wu:fb36b10, wu:fd61a03, zgc:73227
Chromosome Location
Chr.21: 30401780 - 30408763 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212597.2 NP_997762.1 3 - 4 461 72
GenBank mRNA BC059569.1 - 3 - 4 407 72
EH553640.1 - 3 - 4 495 72
EH557479.1 - 1 - 2 490 72

View Full Product Details