Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cul5a
Entrez Gene ID
327215
Gene Name
cullin 5a
Gene Aliases
cul5, wu:fd17d09, wu:fi20h12, xx:11fd17d09, zgc:66185
Chromosome Location
Chr.15: 261839 - 286806 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001328059.1 NP_001314988.1 4 - 5 461 115
GenBank mRNA BC057475.1 - 4 - 5 432 115
BC154320.1 - 4 - 5 425 115

View Full Product Details