Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
yrk
Entrez Gene ID
337571
Gene Name
Yes-related kinase
Gene Aliases
fj84d04, wu:fj84d04, zgc:92165
Chromosome Location
Chr.19: 43392682 - 43432415 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001365553.1 NP_001352482.1 9 - 10 1174 108
NM_212781.2 NP_997946.1 9 - 10 1165 108
GenBank mRNA AY169369.1 - 9 - 10 1125 108
BC081616.1 - 9 - 10 1174 108
FJ205701.1 - 9 - 10 1121 108

View Full Product Details