Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
smtnl
Entrez Gene ID
399690
Gene Name
smoothelin, like
Gene Aliases
wu:fc73c08, zgc:101796
Chromosome Location
Chr.15: 395560 - 424188 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212805.2 NP_997970.1 4 - 5 983 108
GenBank mRNA AB110459.1 - 4 - 5 966 108

View Full Product Details