Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hnrpkl
Entrez Gene ID
406264
Gene Name
heterogeneous nuclear ribonucleoprotein K, like
Gene Aliases
wu:fc19f05, wu:fc48a08, wu:fc84b09, wu:fd16g02, zgc:56206
Chromosome Location
Chr.5: 55603855 - 55625297 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212991.3 NP_998156.3 5 - 6 372 88
XM_009301843.3 XP_009300118.1 5 - 6 426 88
GenBank mRNA BC096867.1 - 5 - 6 352 88
BC164732.1 - 4 - 5 294 88
CT708088.2 - 3 - 4 210 88
EB831522.1 - 5 - 6 367 88

View Full Product Details