Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hnrpkl
Entrez Gene ID
406264
Gene Name
heterogeneous nuclear ribonucleoprotein K, like
Gene Aliases
wu:fc19f05, wu:fc48a08, wu:fc84b09, wu:fd16g02, zgc:56206
Chromosome Location
Chr.5: 55603855 - 55625297 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212991.3 NP_998156.3 7 - 8 557 73
XM_009301843.3 XP_009300118.1 7 - 8 611 73
GenBank mRNA BC096867.1 - 7 - 8 537 73
BC164732.1 - 6 - 7 479 73
CT708088.2 - 5 - 6 395 73

View Full Product Details