Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
plekhj1
Entrez Gene ID
406577
Gene Name
pleckstrin homology domain containing, family J member 1
Gene Aliases
zgc:77177
Chromosome Location
Chr.2: 57848867 - 57871253 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213288.1 NP_998453.1 2 - 3 251 113
GenBank mRNA BC065626.1 - 2 - 3 251 113

View Full Product Details