Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
elovl1b
Entrez Gene ID
406725
Gene Name
ELOVL fatty acid elongase 1b
Gene Aliases
wu:fj32h05, zgc:56567
Chromosome Location
Chr.11: 13094172 - 13152302 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC049330.1 - 3 - 4 504 74

View Full Product Details