Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpn2
Entrez Gene ID
407722
Gene Name
GPN-loop GTPase 2
Gene Aliases
atpbd1b, zgc:92877
Chromosome Location
Chr.16: 33938204 - 33943728 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214801.2 NP_999966.2 2 - 3 532 65
XM_009292455.3 XP_009290730.1 2 - 3 521 65
GenBank mRNA AY576999.1 - 2 - 3 514 65
BC083538.1 - 2 - 3 518 65
BC150450.1 - 2 - 3 509 65
BC152270.1 - 2 - 3 480 65

View Full Product Details