Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpn2
Entrez Gene ID
407722
Gene Name
GPN-loop GTPase 2
Gene Aliases
atpbd1b, zgc:92877
Chromosome Location
Chr.16: 33938204 - 33943728 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214801.2 NP_999966.2 5 - 6 980 80
XM_009292455.3 XP_009290730.1 5 - 6 969 80
GenBank mRNA AY576999.1 - 5 - 6 962 80
BC083538.1 - 5 - 6 966 80
BC150450.1 - 5 - 6 957 80
BC152270.1 - 5 - 6 928 80

View Full Product Details