Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ube2z
Entrez Gene ID
436602
Gene Name
ubiquitin-conjugating enzyme E2Z
Gene Aliases
zgc:92419
Chromosome Location
Chr.3: 23583887 - 23596870 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002330.5 NP_001002330.2 1 - 1 243 155
GenBank mRNA BC159235.1 - 1 - 1 147 155
EB905263.1 - 1 - 1 255 155
EH454696.1 - 1 - 1 246 155

View Full Product Details