Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HTR4
Entrez Gene ID
100034053
Gene Name
5-hydroxytryptamine receptor 4
Gene Aliases
-
Chromosome Location
Chr.14: 28427980 - 28540736 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163982.1 NP_001157454.1 3 - 4 395 112
GenBank mRNA AY647163.1 - 2 - 3 268 112
GU289397.1 - 3 - 4 354 112

View Full Product Details