Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYH1
Entrez Gene ID
791235
Gene Name
myosin heavy chain 1
Gene Aliases
MYHC-2X, MYHC-2d
Chromosome Location
Chr.11: 53325909 - 53350918 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081759.1 NP_001075228.1 2 - 3 269 101
XM_005597036.3 XP_005597093.1 - 373 101
GenBank mRNA AB088366.1 - 2 - 3 269 101

View Full Product Details