Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOS2
Entrez Gene ID
791246
Gene Name
nitric oxide synthase 2
Gene Aliases
INOS
Chromosome Location
Chr.11: 42217303 - 42253897 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081769.1 NP_001075238.1 8 - 9 999 97
XM_023651656.1 XP_023507424.1 - 999 97
XM_023651657.1 XP_023507425.1 - 1385 97
GenBank mRNA AF329377.1 - 2 - 3 241 97
AY027883.1 - 8 - 9 999 97

View Full Product Details