Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LUM
Entrez Gene ID
100009681
Gene Name
lumican
Gene Aliases
-
Chromosome Location
Chr.28: 17947669 - 17954858 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081780.2 NP_001075249.1 2 - 3 956 97
GenBank mRNA AB292110.1 - 2 - 3 956 97
DQ338458.1 - 2 - 3 936 97

View Full Product Details