Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFTPA1
Entrez Gene ID
100033826
Gene Name
surfactant protein A1
Gene Aliases
SFTPA
Chromosome Location
Chr.1: 89779810 - 89784085 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081812.2 NP_001075281.1 1 - 2 194 100
XM_014733440.1 XP_014588926.1 - 433 100
XM_014733441.1 XP_014588927.1 - 353 100
XM_023636212.1 XP_023491980.1 - 255 100
GenBank mRNA AB015963.1 - 1 - 2 195 100

View Full Product Details