Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNT1
Entrez Gene ID
100033893
Gene Name
cyclin T1
Gene Aliases
CYCT1
Chromosome Location
Chr.6: 67467526 - 67491804 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081846.1 NP_001075315.1 3 - 4 372 71
GenBank mRNA AF137509.1 - 3 - 4 372 71
AF190905.1 - 3 - 4 372 71

View Full Product Details