Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAG
Entrez Gene ID
100034171
Gene Name
pregnancy-associated glycoprotein
Gene Aliases
-
Chromosome Location
Chr.12: 24440983 - 24450204 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081943.1 NP_001075412.1 5 - 6 673 81
GenBank mRNA L38511.1 - 5 - 6 673 81

View Full Product Details