Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNASE1
Entrez Gene ID
100034219
Gene Name
deoxyribonuclease 1
Gene Aliases
-
Chromosome Location
Chr.13: 40394643 - 40401610 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082514.2 NP_001075983.1 4 - 5 491 90
XM_023655008.1 XP_023510776.1 - 3557 90
GenBank mRNA AB162819.1 - 4 - 5 491 90

View Full Product Details