Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD2
Entrez Gene ID
100034205
Gene Name
CD2 molecule
Gene Aliases
-
Chromosome Location
Chr.5: 48862488 - 48875526 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085434.1 NP_001078903.1 1 - 2 97 78
GenBank mRNA X69884.1 - - 97 78

View Full Product Details