Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRGPRF
Entrez Gene ID
100049010
Gene Name
MAS related GPR family member F
Gene Aliases
-
Chromosome Location
Chr.12: 31902994 - 31911487 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001093769.1 NP_001087238.1 2 - 3 246 69
XM_005598186.3 XP_005598243.2 - 556 69
XM_005598187.3 XP_005598244.2 - 1951 69
XM_023653789.1 XP_023509557.1 - 437 69
GenBank mRNA AB302196.1 - 2 - 3 246 69

View Full Product Details