Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPIB
Entrez Gene ID
100066834
Gene Name
peptidylprolyl isomerase B
Gene Aliases
-
Chromosome Location
Chr.1: 129306966 - 129312884 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099761.1 NP_001093231.1 1 - 2 142 84
GenBank mRNA EF397503.1 - 1 - 2 142 84

View Full Product Details