Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCAR
Entrez Gene ID
100050205
Gene Name
Fc fragment of IgA, receptor for
Gene Aliases
-
Chromosome Location
Chr.10: 24540964 - 24549072 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099764.1 NP_001093234.1 4 - 5 658 71
GenBank mRNA AB163433.1 - 4 - 5 658 71
AY587560.1 - 4 - 5 647 71
JL623910.1 - 3 - 4 603 71

View Full Product Details