Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD28
Entrez Gene ID
100067829
Gene Name
CD28 molecule
Gene Aliases
-
Chromosome Location
Chr.18: 78478841 - 78505524 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100179.1 NP_001093649.1 1 - 2 57 62
GenBank mRNA EU000412.1 - 1 - 2 57 62

View Full Product Details