Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAGLN
Entrez Gene ID
100062690
Gene Name
transgelin
Gene Aliases
-
Chromosome Location
Chr.7: 25729472 - 25735076 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110134.1 NP_001103604.1 2 - 3 255 65
GenBank mRNA AB364290.1 - 2 - 3 255 65

View Full Product Details