Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LUM
Entrez Gene ID
100009681
Gene Name
lumican
Gene Aliases
-
Chromosome Location
Chr.28: 17947669 - 17954858 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB106281.1 - 1 - 1 54 58
DQ338458.1 - 2 - 2 501 58

View Full Product Details