Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMEL
Entrez Gene ID
100033885
Gene Name
premelanosome protein
Gene Aliases
PMEL17, SILV
Chromosome Location
Chr.6: 74569524 - 74578149 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163889.1 NP_001157361.1 10 - 11 1855 58
XM_005611061.3 XP_005611118.1 - 2082 58
XM_023642149.1 XP_023497917.1 - 2315 58
XM_023642150.1 XP_023497918.1 - 2162 58
GenBank mRNA AF076780.1 - 6 - 7 1275 58

View Full Product Details