Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACTR3
Entrez Gene ID
100033909
Gene Name
actin related protein 3
Gene Aliases
ARP3
Chromosome Location
Chr.18: 16404936 - 16460718 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_023622805.1 XP_023478573.1 - 1431 100
GenBank mRNA AF307853.1 - 2 - 3 166 100

View Full Product Details