Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BMP4
Entrez Gene ID
100034048
Gene Name
bone morphogenetic protein 4
Gene Aliases
-
Chromosome Location
Chr.24: 2243649 - 2250682 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163970.1 NP_001157442.1 4 - 4 1393 89
XM_005605167.2 XP_005605224.1 - 1198 89
XM_005605168.3 XP_005605225.1 - 3126 89
XM_023627706.1 XP_023483474.1 - 1165 89
GenBank mRNA AY500993.1 - 1 - 1 50 89

View Full Product Details