Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSMD4
Entrez Gene ID
100216429
Gene Name
proteasome 26S subunit, non-ATPase 4
Gene Aliases
-
Chromosome Location
Chr.5: 42670571 - 42679371 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142400.1 NP_001135872.1 4 - 5 429 64
XM_005609945.3 XP_005610002.1 - 453 64
XM_005609946.3 XP_005610003.1 - 261 64
XM_023640280.1 XP_023496048.1 - 442 64

View Full Product Details