Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COX10
Entrez Gene ID
100073118
Gene Name
cytochrome c oxidase assembly factor heme A:farnesyltransferase COX10
Gene Aliases
-
Chromosome Location
Chr.11: 56497100 - 56623251 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145274.1 NP_001138746.1 2 - 3 173 85

View Full Product Details