Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PVALB
Entrez Gene ID
100069554
Gene Name
parvalbumin
Gene Aliases
pvalb1
Chromosome Location
Chr.28: 35196997 - 35215434 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159335.1 NP_001152807.1 2 - 3 191 59
XM_005606474.3 XP_005606531.1 - 311 59
XM_014736717.2 XP_014592203.1 - 261 59
XM_014736718.2 XP_014592204.1 - 291 59
GenBank mRNA FM994925.1 - 2 - 3 191 59

View Full Product Details