Assay Design
The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Gene Transcripts
- Gene Symbol
- CD14
- Entrez Gene ID
-
100630219
- Gene Name
- CD14 molecule
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Unknown: 17809 - 19345 on Build EquCab3.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001081927.2 |
|
1 - 2 |
233 |
59 |
GenBank mRNA |
AF200416.2 |
- |
1 - 2 |
100 |
59 |
CD536554.1 |
- |
1 - 2 |
233 |
59 |
- Gene Symbol
- LOC111767791
- Entrez Gene ID
-
111767791
- Gene Name
- monocyte differentiation antigen CD14-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.14: 35562723 - 35564295 on Build EquCab3.0
- Gene Symbol
- LOC111772122
- Entrez Gene ID
-
111772122
- Gene Name
- monocyte differentiation antigen CD14-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Unknown: 31087 - 32659 on Build EquCab3.0
View Full Product Details