Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYB5A
Entrez Gene ID
100052210
Gene Name
cytochrome b5 type A
Gene Aliases
CYB5
Chromosome Location
Chr.8: 92201488 - 92235088 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159732.1 NP_001153204.1 3 - 4 317 81

View Full Product Details