Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL35A
Entrez Gene ID
100034007
Gene Name
ribosomal protein L35a
Gene Aliases
-
Chromosome Location
Chr.19: 36560623 - 36564172 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163880.1 NP_001157352.1 1 - 2 28 56

View Full Product Details