Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MATR3
Entrez Gene ID
100062181
Gene Name
matrin 3
Gene Aliases
-
Chromosome Location
Chr.14: 36628273 - 36658757 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205261.1 NP_001192190.1 1 - 2 164 102
XM_014730313.2 XP_014585799.2 - 153 102

View Full Product Details