Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CIAO2B
Entrez Gene ID
100052873
Gene Name
cytosolic iron-sulfur assembly component 2B
Gene Aliases
FAM96B
Chromosome Location
Chr.3: 18160474 - 18162519 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242528.1 NP_001229457.1 4 - 5 507 56

View Full Product Details