Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH3BP5L
Entrez Gene ID
100049828
Gene Name
SH3 binding domain protein 5 like
Gene Aliases
-
Chromosome Location
Chr.21: 4984244 - 5004465 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242551.1 NP_001229480.1 4 - 5 539 72
XM_005604190.3 XP_005604247.1 - 1117 72
XM_014734839.2 XP_014590325.1 - 813 72
XM_014734840.2 XP_014590326.1 - 658 72
XM_023625240.1 XP_023481008.1 - 925 72
XM_023625241.1 XP_023481009.1 - 549 72

View Full Product Details