Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SELENOI
Entrez Gene ID
100055703
Gene Name
selenoprotein I
Gene Aliases
EPT1, SELI
Chromosome Location
Chr.15: 70744428 - 70791972 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168391.1 NP_001161863.1 1 - 2 204 71

View Full Product Details