Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSRB1
Entrez Gene ID
100068001
Gene Name
methionine sulfoxide reductase B1
Gene Aliases
SEPX1
Chromosome Location
Chr.13: 42053741 - 42057229 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170424.1 NP_001163895.1 3 - 4 372 93

View Full Product Details