Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRIA2
Entrez Gene ID
100069331
Gene Name
glutamate ionotropic receptor AMPA type subunit 2
Gene Aliases
-
Chromosome Location
Chr.2: 76948197 - 77091772 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185087.3 NP_001172016.3 6 - 7 1329 87
NM_001185088.2 NP_001172017.2 6 - 7 1329 87
NM_001185089.2 NP_001172018.2 6 - 7 845 87
XM_005607780.3 XP_005607837.1 - 1418 87
XM_005607783.3 XP_005607840.2 - 1414 87
XM_023626768.1 XP_023482536.1 - 1416 87
XM_023626774.1 XP_023482542.1 - 1416 87
XM_023626782.1 XP_023482550.1 - 1385 87

View Full Product Details