The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204234.1 | NP_001191163.1 | 17 - 18 | 1945 | 90 |
XM_014733464.2 | XP_014588950.1 | - | 2958 | 90 |