Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR27B
- Entrez Gene ID
-
100314892
- Gene Name
- microRNA mir-27b
- Gene Aliases
-
eca-mir-27b
- Chromosome Location
-
Chr.23: 2132383 - 2132455 on Build EquCab3.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_032993.1 |
|
1 - 1 |
38 |
67 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA mir-24-2 |
MIR24-2 |
eca-mir-24-2 |
-452.0
|
Sense |
Chr. - 2131864 - 2131931 on Build EquCab3.0 |
Downstream |
microRNA mir-23b |
MIR23B |
eca-mir-23b |
156.0
|
Sense |
Chr. - 2132611 - 2132684 on Build EquCab3.0 |
View Full Product Details