Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR7-3
- Entrez Gene ID
-
100314894
- Gene Name
- microRNA mir-7-3
- Gene Aliases
-
eca-mir-7-3
- Chromosome Location
-
Chr.23: 6940625 - 6940712 on Build EquCab3.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_032997.1 |
|
1 - 1 |
34 |
82 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
chromosome 23 C9orf64 homolog |
C23H9orf64 |
|
-16.1kb
|
Sense |
Chr. - 6913094 - 6924494 on Build EquCab3.0 |
Downstream |
RecQ mediated genome instability 1 |
RMI1 |
|
10.4kb
|
Anti-sense |
Chr. - 6951134 - 7071459 on Build EquCab3.0 |
View Full Product Details