Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR7-2
- Entrez Gene ID
-
100315008
- Gene Name
- microRNA mir-7-2
- Gene Aliases
-
eca-mir-7-2
- Chromosome Location
-
Chr.7: 3377222 - 3377305 on Build EquCab3.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_032873.1 |
|
1 - 1 |
38 |
66 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
dipeptidyl peptidase 9 |
DPP9 |
|
-59.9kb
|
Anti-sense |
Chr. - 3279638 - 3317330 on Build EquCab3.0 |
Downstream |
fem-1 homolog A |
FEM1A |
|
12.7kb
|
Sense |
Chr. - 3389992 - 3394025 on Build EquCab3.0 |
View Full Product Details