Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIRLET7A-2
- Entrez Gene ID
-
100315027
- Gene Name
- microRNA let-7a-2
- Gene Aliases
-
eca-let-7a-2
- Chromosome Location
-
Chr.28: 43148191 - 43148264 on Build EquCab3.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033060.1 |
|
1 - 1 |
37 |
74 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
translation initiation factor IF-2-like |
LOC111771032 |
|
-41.5kb
|
Anti-sense |
Chr. - 43105386 - 43106683 on Build EquCab3.0 |
Downstream |
peroxisome proliferator activated receptor alpha |
PPARA |
|
32.6kb
|
Sense |
Chr. - 43180883 - 43252049 on Build EquCab3.0 |
View Full Product Details