Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- H19
- Entrez Gene ID
-
100271857
- Gene Name
- H19, imprinted maternally expressed transcript (non-protein coding)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 34334581 - 34336980 on Build EquCab3.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027326.3 |
|
1 - 2 |
1261 |
81 |
GenBank mRNA |
DN511008.1 |
- |
1 - 2 |
77 |
81 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC106781503 |
LOC106781503 |
|
-592.0
|
Anti-sense |
Chr. - 34334439 - 34334918 on Build EquCab3.0 |
Downstream |
insulin like growth factor 2 |
IGF2 |
IGF-II |
102.2kb
|
Sense |
Chr. - 34439224 - 34464997 on Build EquCab3.0 |
View Full Product Details