Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Pichia kudriavzevii
Entrez Gene ID
Pichia kudriavzevii
Gene Name
Candida krusei
Gene Aliases
Pichia kudriavzevii
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length

View Full Product Details