Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCO1
Entrez Gene ID
395346
Gene Name
beta-carotene oxygenase 1
Gene Aliases
BCDO, BCMO1
Chromosome Location
Chr.11: 15458803 - 15472637 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001364902.1 NP_001351831.1 6 - 7 751 138
GenBank mRNA AJ271386.1 - 5 - 6 747 138

View Full Product Details