Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPS14
Entrez Gene ID
424436
Gene Name
mitochondrial ribosomal protein S14
Gene Aliases
-
Chromosome Location
Chr.8: 7476810 - 7479202 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277549.1 NP_001264478.1 2 - 3 215 68
GenBank mRNA BU375572.1 - 2 - 3 215 68
BU456334.1 - 2 - 3 69
BX929470.1 - 2 - 3 191 68
BX933002.1 - 1 - 2 165 68
BX933085.2 - 2 - 3 196 68

View Full Product Details